Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053524.1 | NP_445976.1 | 4 | 332 | ||
XM_008759640.1 | XP_008757862.1 | 4 | 250 | |||
XM_008759641.1 | XP_008757863.1 | 4 | 249 | |||
XM_008759642.1 | XP_008757864.1 | 3 | 222 | |||
XM_008759643.1 | XP_008757865.1 | 4 | 455 | |||
GenBank mRNA | AB044086.1 | - | 4 | 300 | ||
AY027527.1 | - | 4 | 332 |