Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270803.1 | NP_001257732.1 | 2 | 210 | ||
NM_001270804.1 | NP_001257733.1 | 2 | 138 | |||
NM_053529.2 | NP_445981.2 | 2 | 429 | |||
XM_006245738.2 | XP_006245800.1 | 2 | 471 | |||
XM_008767501.1 | XP_008765723.1 | 3 | 1511 | |||
GenBank mRNA | AF251305.1 | - | 2 | 429 | ||
AF251306.1 | - | 2 | 210 | |||
AF251307.1 | - | 2 | 138 |