Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317099.1 | NP_001304028.1 | 2 | 244 | ||
NM_001317103.1 | NP_001304032.1 | 1 | 357 | |||
NM_007034.4 | NP_008965.2 | 1 | 357 | |||
GenBank mRNA | AB209922.1 | - | 1 | 235 | ||
AK313205.1 | - | 1 | 243 | |||
BC034721.1 | - | 1 | 233 | 4340658 | ||
BI831822.1 | - | 2 | 243 | |||
BI910092.1 | - | 1 | 249 | |||
DA354925.1 | - | 1 | 359 | |||
DA473487.1 | - | 1 | 355 | |||
U40992.2 | - | 1 | 250 |