Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053591.2 | NP_446043.1 | 3 | 270 | ||
XM_006255783.2 | XP_006255845.1 | 3 | 265 | |||
GenBank mRNA | BC072476.1 | - | 3 | 270 | 7100683 | |
L07315.1 | - | 3 | 248 | |||
L07316.1 | - | 3 | 248 | |||
M94056.1 | - | 3 | 248 |