Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053633.1 | NP_446085.1 | 2 | 612 | ||
XM_008772857.1 | XP_008771079.1 | 3 | 289 | |||
GenBank mRNA | AB032419.1 | - | 1 | 661 | ||
AB032420.1 | - | 2 | 612 | |||
D83508.1 | - | 2 | 523 | |||
U78102.1 | - | 2 | 598 |