Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145009.1 | NP_001138481.1 | 4 | 841 | ||
NM_007048.5 | NP_008979.3 | 4 | 841 | |||
NM_194441.2 | NP_919423.1 | 4 | 671 | |||
XM_005248833.2 | XP_005248890.1 | 4 | 809 | |||
XM_005248834.3 | XP_005248891.1 | 4 | 836 | |||
GenBank mRNA | AK223474.1 | - | 3 | 622 | ||
AK290193.1 | - | 4 | 838 | |||
AK291162.1 | - | 4 | 671 | |||
AK295720.1 | - | 4 | 828 | |||
AK298652.1 | - | 4 | 570 | |||
BC118586.1 | - | 4 | 802 | 40085016 | ||
BC121800.1 | - | 4 | 802 | 40085015 | ||
BM542639.1 | - | 214 | ||||
U90552.2 | - | 4 | 771 | |||
Y07827.1 | - | 4 | 563 |