Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053654.2 | NP_446106.1 | 2, 3 | 155 | ||
XM_006228365.2 | XP_006228427.1 | 2, 3 | 293 | |||
GenBank mRNA | AF016047.1 | - | 2, 3 | 155 | ||
BC126079.1 | - | 2, 3 | 138 |