Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053683.1 | NP_446135.1 | 6 | 759 | ||
GenBank mRNA | AF152963.1 | - | 6 | 759 |