Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053704.1 | NP_446156.1 | 4 | 402 | ||
XM_006242048.2 | XP_006242110.1 | 4 | 489 | |||
GenBank mRNA | AF372501.1 | - | 4 | 402 | ||
FQ225338.1 | - | 4 | 379 |