Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053715.2 | NP_446167.2 | 1 | 700 | ||
GenBank mRNA | AJ001290.1 | - | 1 | 700 | ||
AY231162.1 | - | 1 | 641 |