Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007055.3 | NP_008986.2 | 3 | 336 | ||
GenBank mRNA | AF021351.1 | - | 4 | 203 | ||
AK300329.1 | - | 3 | 316 | |||
AY091459.1 | - | 3 | 203 | |||
BC041089.1 | - | 3 | 297 | 3915501 | ||
DA103416.1 | - | 3 | 336 |