Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053719.1 | NP_446171.1 | 2 | 277 | ||
GenBank mRNA | AJ009698.1 | - | 2 | 277 | ||
BC061846.1 | - | 2 | 279 | 5621074 | ||
FQ213962.1 | - | 2 | 290 | |||
FQ228768.1 | - | 2 | 290 |