Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLASRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007056.2 | NP_008987.2 | 3 | 253 | ||
NR_103529.1 | 3 | 253 | ||||
XM_011526396.2 | XP_011524698.1 | 3 | 248 | |||
XM_011526397.2 | XP_011524699.1 | 2 | 98 | |||
GenBank mRNA | AF042800.1 | - | 3 | 253 | ||
AK094681.1 | - | 2 | 847 | |||
AY358944.1 | - | 3 | 234 | |||
BC013178.1 | - | 3 | 198 | |||
BC080554.1 | - | 2 | 193 | 6258057 | ||
BC131496.1 | - | 2 | 111 |