Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053729.1 | NP_446181.1 | 10 | 1423 | ||
GenBank mRNA | AY026945.1 | - | 10 | 1423 | ||
BC129088.1 | - | 10 | 1437 | 8362408 | ||
FQ234941.1 | - | 1 | 44 |