Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053755.2 | NP_446207.1 | 6 | 889 | ||
GenBank mRNA | AF314820.1 | - | 3 | 413 | ||
AF337809.1 | - | 6 | 894 |