Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 7 | 1246 | ||
XM_005263862.3 | XP_005263919.1 | 7 | 1535 | |||
XM_005263863.4 | XP_005263920.1 | 7 | 1322 | |||
XM_005263864.3 | XP_005263921.1 | 7 | 1487 | |||
XM_011510507.1 | XP_011508809.1 | 9 | 1377 | |||
XM_011510508.2 | XP_011508810.1 | 7 | 1291 | |||
XM_017003198.1 | XP_016858687.1 | 7 | 1547 | |||
XM_017003199.1 | XP_016858688.1 | 7 | 1500 | |||
XM_017003200.1 | XP_016858689.1 | 7 | 1565 | |||
XM_017003201.1 | XP_016858690.1 | 7 | 1556 | |||
XM_017003202.1 | XP_016858691.1 | 7 | 1591 | |||
GenBank mRNA | AB017114.1 | - | 3 | 374 | ||
AB449889.1 | - | 7 | 1115 | |||
AK292826.1 | - | 5 | 879 | |||
AK300533.1 | - | 7 | 1291 | |||
DB079019.1 | - | 2 | 259 |