Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053819.1 | NP_446271.1 | 2 | 129 | ||
XM_006256608.2 | XP_006256670.1 | 2 | 193 | |||
XM_008773034.1 | XP_008771256.1 | 1 | 103 | |||
GenBank mRNA | AY550026.1 | - | 1 | 118 | ||
BC099821.1 | - | 2 | 127 | 7930800 | ||
FQ219856.1 | - | 2 | 139 | |||
FQ221763.1 | - | 2 | 139 | |||
FQ222271.1 | - | 2 | 139 | |||
FQ222437.1 | - | 2 | 142 | |||
FQ222915.1 | - | 2 | 139 | |||
FQ228400.1 | - | 2 | 138 | |||
FQ230039.1 | - | 2 | 136 | |||
L31883.1 | - | 2 | 129 | |||
U06179.1 | - | 1 | 112 |