Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053835.1 | NP_446287.1 | 6 | 642 | ||
XM_006253595.2 | XP_006253657.1 | 5 | 676 | |||
GenBank mRNA | FQ213983.1 | - | 6 | 732 | ||
FQ229322.1 | - | 5 | 606 | |||
M15883.1 | - | 6 | 642 | |||
M19262.1 | - | 5 | 588 |