Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053836.1 | NP_446288.1 | 2 | 204 | ||
GenBank mRNA | M22899.1 | - | 2 | 204 |