Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053865.1 | NP_446317.1 | 9 | 2763 | ||
XM_008764690.1 | XP_008762912.1 | 7 | 793 | |||
GenBank mRNA | FQ212949.1 | - | 7 | 704 | ||
U17603.1 | - | 7 | 763 | |||
U17604.1 | - | 9 | 2763 |