Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193333.2 | NP_001180262.1 | 5, 6 | 894 | ||
NM_007074.3 | NP_009005.1 | 4, 5 | 776 | |||
XM_011545714.2 | XP_011544016.1 | 4, 5 | 589 | |||
XM_017022885.1 | XP_016878374.1 | 4, 5 | 764 | |||
XM_017022886.1 | XP_016878375.1 | 4, 5 | 586 | |||
GenBank mRNA | AB209221.1 | - | 2 | 2721 | ||
AF495470.1 | - | 4, 5 | 581 | |||
AK097964.1 | - | 1 | 365 | |||
AK123401.1 | - | 2 | 1020 | |||
AK296207.1 | - | 4 | 670 | |||
AK314714.1 | - | 5, 6 | 695 | |||
BC110374.1 | - | 4, 5 | 593 | 5733853 | ||
BC126385.1 | - | 4, 5 | 480 | 8992101 | ||
BC126387.1 | - | 4, 5 | 480 | 8992103 | ||
D44497.1 | - | 4, 5 | 559 | |||
GQ214031.1 | - | 382 | ||||
U34690.1 | - | 4, 5 | 551 | |||
X89109.1 | - | 4, 5 | 552 |