Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053893.3 | NP_446345.3 | 4 | 1187 | ||
GenBank mRNA | U52825.1 | - | 4 | 1190 | ||
U73184.1 | - | 4 | 1255 |