Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029896.1 | NP_001025067.1 | 5 | 447 | ||
NM_007075.3 | NP_009006.2 | 6 | 717 | |||
GenBank mRNA | AJ005897.1 | - | 5 | 403 | ||
AK023344.1 | - | 3 | 1425 | |||
AK225122.1 | - | 5 | 426 | |||
AK315717.1 | - | 5 | 434 | |||
AY691428.1 | - | 4 | 275 | |||
BC000464.2 | - | 5 | 441 | 2822742 | ||
BC003037.1 | - | 5 | 441 | 2822742 | ||
BC009027.1 | - | 2 | 71 | |||
BC069206.1 | - | 6 | 717 | 5801768 | ||
BQ068083.1 | - | 5 | 447 | |||
CR456725.1 | - | 4 | 275 |