Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053949.1 | NP_446401.1 | 10, 11 | 2597 | ||
XM_006235867.2 | XP_006235929.1 | 10, 11 | 2730 | |||
XM_008762630.1 | XP_008760852.1 | 6, 7 | 1918 | |||
GenBank mRNA | AY669863.1 | - | 6, 7 | 1571 | ||
Z96106.1 | - | 10, 11 | 2597 |