Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007080.2 | NP_009011.1 | 3, 4 | 346 | ||
XM_017007672.1 | XP_016863161.1 | 3, 4 | 321 | |||
XM_017007673.1 | XP_016863162.1 | 3, 4 | 646 | |||
GenBank mRNA | AF182292.1 | - | 3, 4 | 288 | ||
AJ238098.1 | - | 3, 4 | 288 | |||
AK312126.1 | - | 3, 4 | 281 | |||
BC016026.1 | - | 3, 4 | 300 | 3910698 | ||
BC019894.1 | - | 3, 4 | 926 | |||
DB465201.1 | - | 3, 4 | 347 |