Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053971.3 | NP_446423.2 | 1 | 235 | ||
XM_006249377.1 | XP_006249439.1 | 2 | 254 | |||
XM_006249378.2 | XP_006249440.1 | 2 | 288 | |||
GenBank mRNA | BC061784.1 | - | 1 | 231 | ||
BC078761.1 | - | 1 | 238 | 7123086 | ||
BC097295.1 | - | 1 | 239 | 7446680 | ||
FQ222362.1 | - | 2 | 251 | |||
FQ229484.1 | - | 2 | 253 | |||
FQ229943.1 | - | 2 | 252 | |||
FQ234880.1 | - | 2 | 252 | |||
X87107.1 | - | 2 | 249 |
Gene Symbol: LOC681117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760839.1 | XP_008759061.1 | 4 | 845 |