Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053990.1 | NP_446442.1 | 3 | 247 | ||
XM_006254854.2 | XP_006254916.1 | 3 | 1486 | |||
XM_006254855.2 | XP_006254917.1 | 3 | 1486 | |||
XM_008772080.1 | XP_008770302.1 | 3 | 1486 | |||
XM_008772081.1 | XP_008770303.1 | 3 | 1486 | |||
GenBank mRNA | BC078758.1 | - | 3 | 286 | 7132485 | |
X58828.1 | - | 3 | 247 |