Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057114.1 | NP_476455.1 | 2 | 152 | ||
GenBank mRNA | BC058450.1 | - | 2 | 172 | 6920987 | |
BC088118.1 | - | 2 | 150 | 7324630 | ||
D30035.1 | - | 2 | 152 | |||
FQ219859.1 | - | 2 | 169 |