Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080694.1 | NP_542425.1 | 1 | 86 | ||
XM_008758749.1 | XP_008756971.1 | 1 | 1166 | |||
GenBank mRNA | AF361343.1 | - | 1 | 86 | ||
AF361344.1 | - | 1 | 86 |