Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080696.2 | NP_542427.1 | 4 | 1246 | ||
GenBank mRNA | AF361346.1 | - | 4 | 1246 |