Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080908.1 | NP_543184.1 | 12 | 1299 | ||
XM_008771422.1 | XP_008769644.1 | 12 | 1384 | |||
GenBank mRNA | AY059414.1 | - | 12 | 1299 |