Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130749.1 | NP_570105.1 | 10 | 996 | ||
XM_006240563.2 | XP_006240625.1 | 10 | 1130 | |||
XM_006240564.2 | XP_006240626.1 | 10 | 1435 | |||
XM_006240565.2 | XP_006240627.1 | 10 | 1130 | |||
XM_006240566.2 | XP_006240628.1 | 10 | 1130 | |||
XM_006240567.2 | XP_006240629.1 | 10 | 1130 | |||
XM_006240568.2 | XP_006240630.1 | 10 | 1130 | |||
XM_006240569.2 | XP_006240631.1 | 10 | 1130 | |||
XM_006240570.2 | XP_006240632.1 | 10 | 1130 | |||
XM_006240571.2 | XP_006240633.1 | 10 | 1130 | |||
GenBank mRNA | AF465412.1 | - | 10 | 996 | ||
FQ214295.1 | - | 6 | 523 | |||
FQ235189.1 | - | 9 | 700 |