Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130828.2 | NP_570841.2 | 8 | 1174 | ||
GenBank mRNA | AB091379.1 | - | 8 | 1113 | ||
AF400653.1 | - | 6 | 759 | |||
AY039651.1 | - | 8 | 1174 | |||
BC098649.1 | - | 8 | 1156 | 7127895 |