Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131906.1 | NP_571981.1 | 15 | 2914 | ||
XM_008763351.1 | XP_008761573.1 | 15 | 3002 | |||
GenBank mRNA | U88036.1 | - | 15 | 2914 |