Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270807.1 | NP_001257736.1 | 5 | 847 | ||
NM_001270808.1 | NP_001257737.1 | 5 | 896 | |||
NM_001270809.1 | NP_001257738.1 | 5 | 797 | |||
NM_001270810.1 | NP_001257739.1 | 5 | 1483 | |||
NM_001270811.1 | NP_001257740.1 | 5 | 1833 | |||
NM_133386.3 | NP_596877.2 | 4 | 2087 | |||
XM_006247787.2 | XP_006247849.1 | 5 | 1167 | |||
XM_006247788.2 | XP_006247850.1 | 5 | 678 | |||
GenBank mRNA | AB049571.1 | - | 5 | 895 | ||
AB049572.1 | - | 5 | 847 | |||
AB049573.1 | - | 5 | 794 | |||
AB049574.1 | - | 5 | 1453 | |||
AB049575.1 | - | 4 | 1600 | |||
BC081738.1 | - | 4 | 2052 | 7132896 | ||
DQ486894.1 | - | 5 | 635 |