Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133393.2 | NP_596884.1 | 8 | 1769 | ||
GenBank mRNA | BC070933.1 | - | 8 | 1769 | 7097855 |