Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133401.1 | NP_596892.1 | 4, 5 | 345 | ||
XM_006235994.2 | XP_006236056.1 | 6, 7 | 1170 | |||
GenBank mRNA | AF257746.1 | - | 4, 5 | 345 | ||
AY582535.1 | - | 3, 4 | 260 |