Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133406.1 | NP_596897.1 | 5 | 750 | ||
XM_006227856.2 | XP_006227918.1 | 5 | 1465 | |||
XM_006227857.2 | XP_006227919.1 | 5 | 1119 | |||
GenBank mRNA | AB067572.1 | - | 5 | 750 | ||
BC086992.1 | - | 5 | 669 | 7191675 |