Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007148.4 | NP_009079.2 | 6 | 1012 | ||
XM_006721571.3 | XP_006721634.1 | 6 | 5020 | |||
XM_006721572.3 | XP_006721635.1 | 6 | 5018 | |||
XM_006721573.3 | XP_006721636.1 | 6 | 753 | |||
GenBank mRNA | AB026054.1 | - | 6 | 866 | ||
AK226052.1 | - | 6 | 952 | |||
AK293508.1 | - | 4 | 621 | |||
AK313844.1 | - | 6 | 958 | |||
BC053989.1 | - | 6 | 927 | 5922474 |