Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133514.1 | NP_598198.1 | 7 | 1011 | ||
GenBank mRNA | M65253.1 | - | 7 | 1011 | ||
X05083.1 | - | 7 | 1008 |