Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19090
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
20
1872
NM_001178107.1
NP_001171578.1
19
1785
NM_001178108.1
NP_001171579.1
19
1779
NM_001178109.1
NP_001171580.1
18
1689
NM_001178110.1
NP_001171581.1
17
1599
NM_001178113.1
NP_001171584.1
11
1329
NM_001178115.1
NP_001171586.1
5
680
NM_007150.3
NP_009081.2
19
1776
XM_005274730.2
XP_005274787.1
21
1979
XM_005274731.2
XP_005274788.1
21
1976
XM_005274735.2
XP_005274792.1
20
1889
XM_005274737.2
XP_005274794.1
20
1883
XM_005274738.2
XP_005274795.1
20
1880
XM_005274740.2
XP_005274797.1
19
1793
XM_005274741.2
XP_005274798.1
19
1793
XM_005274742.2
XP_005274799.1
19
1790
XM_005274744.2
XP_005274801.1
18
1706
XM_005274745.2
XP_005274802.1
18
1703
XM_005274746.3
XP_005274803.1
17
1622
XM_011531194.1
XP_011529496.1
19
1784
XM_011531195.1
XP_011529497.1
20
1781
XM_017029821.1
XP_016885310.1
22
2111
XM_017029822.1
XP_016885311.1
23
2131
XM_017029823.1
XP_016885312.1
22
2108
XM_017029824.1
XP_016885313.1
21
2027
XM_017029825.1
XP_016885314.1
21
2024
XM_017029826.1
XP_016885315.1
21
2021
XM_017029827.1
XP_016885316.1
21
2018
XM_017029828.1
XP_016885317.1
21
2015
XM_017029829.1
XP_016885318.1
20
1934
XM_017029830.1
XP_016885319.1
20
1928
XM_017029831.1
XP_016885320.1
20
1925
XM_017029832.1
XP_016885321.1
20
1916
XM_017029833.1
XP_016885322.1
19
1850
XM_017029834.1
XP_016885323.1
19
1838
XM_017029835.1
XP_016885324.1
19
1799
GenBank mRNA
AK056517.1
-
8
835
AK095258.1
-
5
652
AK296967.1
-
19
1779
AK301530.1
-
11
1329
AK304323.1
-
15
1342
AY997296.1
-
19
1765
BX537525.1
-
5
2354
BX640940.1
-
11
1076
DQ508022.1
-
19
1728
DQ508023.1
-
17
1551
DQ508024.1
-
18
1641
DQ508025.1
-
20
1824
DQ508026.1
-
19
1737
Y09538.1
-
11
1057
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001109043, NM_009549, U46687, EF494744, AK014497, and others.
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline