Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133544.1 | NP_598228.1 | 2 | 194 | ||
GenBank mRNA | AJ271158.1 | - | 2 | 194 | ||
FQ217274.1 | - | 3 | 201 | |||
FQ217509.1 | - | 3 | 254 | |||
FQ217783.1 | - | 3 | 253 | |||
FQ221212.1 | - | 3 | 252 | |||
FQ223588.1 | - | 3 | 249 | |||
FQ223759.1 | - | 3 | 201 |
Gene Symbol: LOC103693430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768219.1 | XP_008766441.1 | 1 | 229 |