Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304421.2 | NP_001291350.1 | 10 | 1522 | ||
NM_007159.4 | NP_009090.2 | 10 | 1522 | |||
XM_005265465.3 | XP_005265522.1 | 11 | 2358 | |||
XM_005265466.3 | XP_005265523.1 | 11 | 2352 | |||
XM_005265470.3 | XP_005265527.1 | 11 | 2358 | |||
XM_005265471.3 | XP_005265528.1 | 11 | 2352 | |||
XM_006713326.2 | XP_006713389.1 | 11 | 2352 | |||
XM_011534104.1 | XP_011532406.1 | 11 | 2358 | |||
XM_011534105.1 | XP_011532407.1 | 11 | 2358 | |||
XM_017007151.1 | XP_016862640.1 | 11 | 2358 | |||
XM_017007152.1 | XP_016862641.1 | 11 | 2352 | |||
XM_017007158.1 | XP_016862647.1 | 11 | 2358 | |||
XM_017007160.1 | XP_016862649.1 | 11 | 2352 | |||
XM_017007164.1 | XP_016862653.1 | 11 | 2352 | |||
XM_017007167.1 | XP_016862656.1 | 11 | 2358 | |||
XM_017007168.1 | XP_016862657.1 | 11 | 2352 | |||
GenBank mRNA | AB046821.1 | - | 8 | 763 | ||
AF100750.1 | - | 1 | 122 | |||
BC114627.1 | - | 10 | 1220 | 40081892 |