Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133594.2 | NP_598278.1 | 2 | 304 | ||
XM_008768397.1 | XP_008766619.1 | 3 | 444 | |||
GenBank mRNA | BC058446.1 | - | 267 | 6921523 | ||
BC078746.1 | - | 2 | 238 | 7125132 | ||
CF976332.1 | - | 308 | ||||
FQ221996.1 | - | 271 | ||||
FQ225379.1 | - | 246 | ||||
FQ225921.1 | - | 262 | ||||
FQ225989.1 | - | 262 | ||||
FQ230134.1 | - | 273 | ||||
FQ235035.1 | - | 2 | 185 | |||
L79949.1 | - | 2 | 251 |
Gene Symbol: Sumo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729458.4 | XP_002729504.1 | 1 | 241 |