Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133594.2 | NP_598278.1 | 4 | 497 | ||
XM_008768397.1 | XP_008766619.1 | 5 | 637 | |||
GenBank mRNA | BC058446.1 | - | 460 | 6921523 | ||
BC078746.1 | - | 4 | 431 | 7125132 | ||
CF976332.1 | - | 501 | ||||
FQ221996.1 | - | 464 | ||||
FQ225379.1 | - | 439 | ||||
FQ225921.1 | - | 455 | ||||
FQ225989.1 | - | 455 | ||||
FQ230134.1 | - | 466 | ||||
L79949.1 | - | 4 | 444 |