Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133605.1 | NP_598289.1 | 8 | 595 | ||
XM_008770486.1 | XP_008768708.1 | 8 | 667 | |||
XM_008770487.1 | XP_008768709.1 | 8 | 667 | |||
XM_008770488.1 | XP_008768710.1 | 8 | 666 | |||
XM_008770489.1 | XP_008768711.1 | 8 | 666 | |||
XM_008770490.1 | XP_008768712.1 | 8 | 666 | |||
XM_008770491.1 | XP_008768713.1 | 8 | 666 | |||
XM_008770492.1 | XP_008768714.1 | 8 | 666 | |||
XM_008770493.1 | XP_008768715.1 | 8 | 665 | |||
XM_008770494.1 | XP_008768716.1 | 8 | 665 | |||
XM_008770495.1 | XP_008768717.1 | 8 | 665 | |||
XM_008770496.1 | XP_008768718.1 | 8 | 665 | |||
XM_008770497.1 | XP_008768719.1 | 8 | 664 | |||
XM_008770498.1 | XP_008768720.1 | 8 | 664 | |||
XM_008770499.1 | XP_008768721.1 | 8 | 664 | |||
GenBank mRNA | J04063.1 | - | 8 | 595 | ||
S71571.1 | - | 8 | 560 |
Gene Symbol: NEWGENE_621802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770639.1 | XP_008768861.1 | 7 | 620 |