Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133624.2 | NP_598308.1 | 2 | 333 | ||
GenBank mRNA | BC086991.1 | - | 2 | 295 | 7191761 | |
M80367.1 | - | 2 | 334 |
Gene Symbol: Gbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233425.2 | XP_006233487.1 | 2 | 308 | ||
XM_008761538.1 | XP_008759760.1 | 3 | 503 |
Gene Symbol: Gbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749410.3 | XP_003749458.2 | 2 | 299 | ||
XM_006233426.2 | XP_006233488.1 | 2 | 299 |
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108569.3 | NP_001102039.2 | 2 | 399 | ||
GenBank mRNA | CB615696.1 | - | 2 | 412 | ||
FM048046.1 | - | 2 | 399 |