Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 2 | 471 | ||
GenBank mRNA | AJ298278.1 | - | 2 | 471 | ||
BC083176.1 | - | 2 | 715 | 7097075 |