Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134461.1 | NP_604456.1 | 10 | 1456 | ||
XM_006229729.2 | XP_006229791.2 | 10 | 1693 | |||
XM_006229730.2 | XP_006229792.1 | 10 | 1585 | |||
XM_006229731.2 | XP_006229793.1 | 10 | 1829 | |||
XM_008759673.1 | XP_008757895.1 | 9 | 1620 | |||
GenBank mRNA | AF484958.1 | - | 10 | 1456 | ||
FQ223532.1 | - | 10 | 1550 |