Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134465.2 | NP_604460.2 | 8 | 1215 | ||
XM_008769915.1 | XP_008768137.1 | 8 | 1173 | |||
XM_008769916.1 | XP_008768138.1 | 8 | 1198 | |||
GenBank mRNA | AF404510.1 | - | 8 | 1158 | ||
BC078740.1 | - | 8 | 1215 | 7108137 |